Would you like to log in again?
Saber Tooth Trimmable Ceramic Flap Discs, Weiler®
Supplier: Weiler
804-50121
300085-688EA 20.71 USD InStock
300085-688
Saber Tooth Trimmable Ceramic Flap Discs, Weiler®
Abrasives
  • Fast cut and long life due to the aggressive self sharpening ceramic grain
  • Specially formulated grinding aid prevents dulling and glazing of grain and allows for "cool cutting" protecting high value parts from heat discoloration
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty