Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proto® 440SS Top Chests, 18 in Deep, Red, Stanley® Products

Supplier: Stanley
Proto®
577-442719-6RD
300025-126EA 1251.52 USD InStock
300025-126
Proto® 440SS Top Chests, 18 in Deep, Red, Stanley® Products
Furniture Storage Cabinets
Smooth, sleek and durable paint finish.

  • Maximum weight capacity for each drawer is 100 lbs.
  • Aluminum extruded drawer pulls
  • 100% full extension drawers
  • Recessed drawer pulls
  • 0.040" drawer front and body
  • Heavy duty key

Drawer dimensions:
1 drawer: 23¹/₄W×16¹/₈D×3¹⁵/₁₆" H
5 drawers: 23¹/₄W×16¹/₈D×1⁷/₈" H

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty