Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Reagent alcohol 190° Proof

Supplier: Decon Labs
Danger
Synonyms: Reagent alcohol

12R1901
76048-924CS 232.96 USD InStock
76048-924
Reagent alcohol 190° Proof
Alcohol
Reagent alcohol 190 proof is a specially denatured ethanol product used in the clinical histology lab.

  • Denatured according to formula 3A

Appearance: Clear
Odor: Mild


Boiling Pt: 78 °C
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty