Would you like to log in again?
DiscoveryPak™ Epigenetic Modulators Set I, Biovision
Supplier: BioVision
AB286234-1KIT
76022-130EA 1853.95 USD InStock
76022-130
DiscoveryPak™ Epigenetic Modulators Set I, Biovision
Assays Cellular Assays
DiscoveryPak™ Epigenetic Modulators Set I is a convenient set of ten (10) small molecule modulators useful in epigenetics research
Each set contains 5 mg of Anacardic acid (Cat. No. 1849-5), 5 mg of BIX01294 (Cat. No. 1678-5), 250 µg of 3-Deazaneplaocin A (Cat. No. 2060-250), 1 mg of EPZ-5676 (Cat. No. 2382-1), 1 mg of GSK-J1 sodium salt (Cat. No. 2260-1), 1 mg of GSK-126 (Cat. No. 2282-1), 5 mg of JIB-04 (Cat. No. 2474-5), 1 mg of (+)-JQ1 (Cat. No. 2070-1), 10 mg of RG 108 (Cat. No. 1679-10), and 1 mg of Trichostatin A (Cat. No. 1606-1)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty