Would you like to log in again?
Quick Connects, Western Enterprises
Supplier: Western Enterprises
312-QDB33 312-QDB31 312-QDB21 312-QDB11 312-QDB12 312-QDB32 312-QDB22 312-QDB34
300008-005EA 49.04 USD InStock
300008-005 300066-914 300008-006 300066-910 300008-002 300066-912 300008-003 300066-908
Quick Connects, Western Enterprises
Welding Supplies
  • Brass construction provides long service life
  • Non-interchangeable oxygen and fuel gas connectors
  • Instant closure on reverse flow check valves
  • Spring-loaded locking mechanism responds to fingertip action and achieves a gas-tight connection
  • Double seal socket connection on second seal
  • Color-coding for gas service type provides additional safety
Certifications: Exceed CGA Flow Specifications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty