Would you like to log in again?
Check Valves, Western Enterprises
Supplier: Western Enterprises
312-CV-7R 312-CV-8L 312-WE-63 312-CV-A10 312-CV-32R 312-WE-60
300067-082EA 51.26 USD InStock
300067-082 300007-973 300007-974 300008-023 300066-730 300066-744
Check Valves, Western Enterprises
Welding Supplies
  • Machined high quality brass with stainless steel operating spring and valve disc
  • Prevents reverse flow at point of origin
  • Easy to install
Certifications: Meets or Exceeds CGA and OSHA Requirements.; UL Listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty