Would you like to log in again?
Inline Check Valves, Western Enterprises
Supplier: Western Enterprises
312-CV-5 312-CV-6 312-CV-4
300066-738EA 43.89 USD InStock
300066-738 300066-732 300066-740
Inline Check Valves, Western Enterprises
Welding Supplies
Check Valves are appropriate for applications that require higher operating pressures, up to 200psig
Compact design and low/open/close pressures: 3oz. to open valve and 10oz., or reverse pressure for positive valve closing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty