Would you like to log in again?
Check Valves, Torch Model A Size, Western Enterprises
Supplier: Western Enterprises
312-CV-A9
300066-746EA 266.19 USD InStock
300066-746
Check Valves, Torch Model A Size, Western Enterprises
Welding Supplies
One-piece, compact unit designed for A-Size applications
Mounts to torch to prevent reverse flow at point of origin.
Certifications: Meets or Exceeds CGA and OSHA Requirements.; UL Listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty