Would you like to log in again?
Flashback Arrestor Assemblies, Western Enterprises
Supplier: Western Enterprises
312-FA-510CV
300066-810EA 200.71 USD InStock
300066-810
Flashback Arrestor Assemblies, Western Enterprises
Welding Supplies
Designed for installation on acetylene cylinders to prevent flashbacks into the cylinder
  • Extinguish flashes or flames before they reach the cylinder
  • Rated flow capacity is 40 SCFH with a 5 psig pressure drop
  • Includes check valve
Flashback Arrestor Assemblies can be used in any lead not exceeding 3/8" pipe in size which connects an acetylene cylinder to a regulator, manifold, or other equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty