Would you like to log in again?
Safemate Quick Connect Sets with Flash Arrestors, Western Enterprises
Supplier: Western Enterprises
312-QDF30 312-QDF10
300066-926EA 139.43 USD InStock
300066-926 300066-928
Safemate Quick Connect Sets with Flash Arrestors, Western Enterprises
Welding Supplies
  • Color-coded for gas service to provide added safety
  • Internal check valves designed to prevent reverse flow and accidental mixing of oxygen and fuel gases
  • Torch and regulator models provide downstream and upstream protection
  • 10" maximum cutting capacity
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty