Would you like to log in again?
Proto® Parallel Jaw Lock Ring Plier, ORS Nasco
Supplier: ORS Nasco
577-251G
300024-188EA 89.74 USD InStock
300024-188
Proto® Parallel Jaw Lock Ring Plier, ORS Nasco
Pliers
For the removal of lock snap rings used on bearing shafts.
  • Compound leverage simplifies removal
  • Indented jaw tips hold ring ends and prevent breakage
  • Plastisol grips for added comfort
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty