Would you like to log in again?
Diaphragm Pumps, Ingersoll-Rand
Supplier: Ingersoll-Rand
5011130987
300054-890EA 831.4 USD InStock
300054-890
Diaphragm Pumps, Ingersoll-Rand
Pumps Vacuum Pumps
Pumps feature a patented "unbalanced" air valve design that avoids stall-out, even under low air inlet pressures
  • PTFE diaphragm
  • Polypropylene body
  • For use with all materials, except for highly abrasive substances
Utilize bolted fasteners for leak-tight integrity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty