Would you like to log in again?
Human Beta-Amyloid (1-40),Biotin
Supplier: AnaSpec
AS-23512-01 AS-23512
103003-538EA 209 USD InStock
103003-538 103003-536
Human Beta-Amyloid (1-40),Biotin
Proteins and Peptides
Aß (1-40) together with Aß (1-42) are two major C-terminal variants of the Aß protein constituting the majority of Aßs. These undergo post-secretory aggregation and deposition in the Alzheimer’s disease brain. This peptide is biotinylated at its N-terminus.
  • Peptide Content: ≥ 60%
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty