Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Beta-Amyloid (3-40)

Supplier: AnaSpec
AS-61029
103006-264EA 173.92 USD InStock
103006-264
Human Beta-Amyloid (3-40)
Proteins and Peptides
This peptide is beta-amyloid (1-40) N-terminally truncated. It is the non-pyroglatamate form of beta-Amyloid (3-40). N-terminally truncated pyroglutamate-modified beta-Amyloid forms such as Aß(3-40) and Aß (11- 40) have been described as major compounds in the senile plaques. Pyro-Glu modified beta-Amyloid forms are more resistant to degradation, show higher toxicity and have increased aggregation propensity compared to non-modified beta-Amyloid.
Sequence: EFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVV
Molecular Weight: 4143.7 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty