Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human;Mouse;Rat Beta-Amyloid (25-35) HCl

Supplier: AnaSpec
AS-23212
102999-434EA 559.77 USD InStock
102999-434
Human;Mouse;Rat Beta-Amyloid (25-35) HCl
Proteins and Peptides
All the HCl salt forms of Aß (1-40), Aß (25-35) and D-Ser26 Aß (25-35), take the ß-structure within a few hours in PBS. They form fibrils, exert toxic effects on hippocampal cultured neurons and suppress MTT reduction activity of non-neuronal (HeLa) cells without cytotoxicity.
Sequence: GSNKGAIIGLM - HCl
Molecular Weight: 1060.3+36.5 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty