Would you like to log in again?
Nitrotyrosine IP Kit, Cayman Chemical Company
Supplier: Cayman Chemical
601220-1
76170-296EA 572.8 USD InStock
76170-296
Nitrotyrosine IP Kit, Cayman Chemical Company
Assays Cellular Assays
For the capture and pulldown of nitrated proteins from cell and tissue lysates.
  • Determine change in tyrosine nitration in treated versus non-treated samples
  • Demonstrate target proteins are nitrated in vitro
  • Identify and characterize nitrated proteins by Western blot and proteomic analysis
Cayman’s Nitrotyrosine IP Kit allows for the capture and pulldown of nitrated proteins from cell lysates using a sorbent coupled with a nitrotyrosine monoclonal antibody. The nitrated proteins can be eluted from the resin, loaded to an SDS-PAGE gel, and probed using the included Nitrotyrosine Polyclonal Antibody or a user supplied target specific antibody. A positive control sample assures that the assay is working.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty