Would you like to log in again?
Diaphragm Tilt Valve Balloon Inflators, Western Enterprises
Supplier: Western Enterprises
312-RPB-5HG
300066-960EA 157.76 USD InStock
300066-960
Diaphragm Tilt Valve Balloon Inflators, Western Enterprises
Welding Supplies
  • Easily adjusted to increase outlet pressures to 125 psi
  • Equipped with an internal safety relief device
  • Includes filters to protect against contaminant and dirt damage
  • Ideal for use with various latex sizing machines
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty