Would you like to log in again?
Economy Balloon Inflators with Gauge, Western Enterprises
Supplier: Western Enterprises
312-R125G
300008-007EA 83.96 USD InStock
300008-007
Economy Balloon Inflators with Gauge, Western Enterprises
Welding Supplies
  • Horizontal one-piece design with all brass piston construction
  • Replaceable Viton® O-ring
  • Hand tight design provides leak-free connection
  • Replaceable rubber tilt valve
  • Easy-to-read contents gauge with rubber protective cover
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty