Would you like to log in again?
Flow Gauge Regulators, Argon, Western Enterprises
Supplier: Western Enterprises
312-REB-3-FGD 312-REB-3-FG
300066-936EA 309.23 USD InStock
300066-936 300066-934
Flow Gauge Regulators, Argon, Western Enterprises
Welding Supplies
Dual scale flow gauge calibrated for Argon/Carbon Dioxide services.
  • Durable Neoprene diaphragm maintains consistent flow rates
  • Protective inlet filtering system extends regulator life
  • KEL-F® seat assembly
Regulator 300066-936 includes a 6' Neoprene hose with fittings.
Certifications: UL Listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty