Would you like to log in again?
Replacement Tilt Valve, Western Enterprises
Supplier: Western Enterprises
312-RPB-5-1V
300008-009EA 14.05 USD InStock
300008-009
Replacement Tilt Valve, Western Enterprises
Welding Supplies
Replacement tilt valve for balloon fillers
Brass construction with durable rubber tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty