Would you like to log in again?
RHP High Inlet/Delivery Pressure Regulators, Compressed Air, Western Enterprises
Supplier: Western Enterprises
312-RHP-2-4
300066-948EA 905.65 USD InStock
300066-948
RHP High Inlet/Delivery Pressure Regulators, Compressed Air, Western Enterprises
Welding Supplies
Consistent delivery pressure, minimizes fluctuation over a wide rage of inlet pressures
  • Dual safety relief valves protect against over-pressurization
  • Self re-seating pressure relief valve
  • Replaceable Delrin® thrust bushing for easy adjustment
  • Nickel chrome plated zinc bonnet
  • Comp1™ pressure compensated design
  • KEL-F® seat assembly
  • Dual inlet filtering system
Use for high pressure cylinders up to 6000psig with applications that require high flow rates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty