Would you like to log in again?
RP Series F.R.O.G. Preset Flow Regulators, Western Enterprises
Supplier: Western Enterprises
312-RP30580 312-RP22320
300066-954EA 83.58 USD InStock
300066-954 300066-956
RP Series F.R.O.G. Preset Flow Regulators, Western Enterprises
Welding Supplies
Easy-to-use preset flow regulators require no adjustment
  • Protective inlet filter extends reuglator life
  • Internal safety relief quards against over-pressurization
  • Dependable brass piston with PTFE seat
Designed for supplying a preset flow of inert shielding gas for M.I.G. welding applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty