Would you like to log in again?
RPB-3 Gas Savers, Argon/CO₂, Western Enterprises
Supplier: Western Enterprises
312-RPB-3
300066-958EA 235.57 USD InStock
300066-958
RPB-3 Gas Savers, Argon/CO₂, Western Enterprises
Welding Supplies
Chrome plated zinc bonnet
  • Machined aluminum body
  • Neoprene diaphragm
  • 2" nominal width
Factory set at 4 PSIG but easily adjustable to meet a range of requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty