Would you like to log in again?
Bobcat Drive Mandrel, Weiler®
Supplier: Weiler
804-07766
300010-134EA 24.51 USD InStock
300010-134
Bobcat Drive Mandrel, Weiler®
Abrasives
For mounting 2 and 3" BobCats on tools with 1/4" collets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty