Would you like to log in again?
Tweezers WA1 Handle with 52-STD Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
10004-S52-SM TIP-52-STD
103302-602EA 67.19 USD InStock
103302-602 103301-882
Tweezers WA1 Handle with 52-STD Tips, Electron Microscopy Sciences
Forceps
These tweezers have an anti-magnetic WA1 handle with replaceable 52-STD tips.
  • Hand made of the highest quality steels and alloys
  • Tips attach with screws
  • High precision and perfectly finished to suit the application
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as inox.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty