Would you like to log in again?
Pierce™ PEGylated BM (bismaleimido) Crosslinkers, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
22337 22336 22337 22336
PI22336EA 245.54 USD InStock
CAPI22337 PI22336 PI22337 CAPI22336
Pierce™ PEGylated BM (bismaleimido) Crosslinkers, Thermo Scientific
Protein Linking and Labeling Systems
Cross linkers are used for the identification of near-neighbor protein relationships and ligand-receptor interactions
Thermo Scientific Pierce BM(PEG)n is a bismaleimide-activated PEG compound for crosslinking between sulfhydryl (—SH) groups in proteins and other thiol molecules.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty