Would you like to log in again?
Chromolith® Prep HPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
Chromolith®
1.25251.0001 1.25260.0001 1.25260.0001 1.25252.0001 1.25252.0001 1.25251.0001 1.25261.0001 1.25261.0001
10143-120EA 9290.91 USD InStock
10143-120 CA1.25251.0001 CA1.25261.0001 EM1.25260.0001 EM1.25261.0001 CA1.25252.0001 10143-118 CA1.25260.0001
Chromolith® Prep HPLC Columns, MilliporeSigma
Chromatography Columns
Chromolith® Prep monolithic stationary phases are new ultrapure silica phases which gain their special properties from a bimodal pore structure.
The large macropores, with a pore diameter ranging from 3 µm, form a dense network of pores and allow a high flow rate due to a low resistance factor. The resulting excellent accessibility of the mesopores (total porosity >80%) assures fast adsorption and desorption kinetics because of the short diffusion length inside the pores. This results in dramatically reduced separation times and thus greatly increased productivity.
To operate a monolithic column it is necessary to ensure that no flow-through occurs in the wall region. Therefore the monolith is cladded with a polymeric material (special PEEK) and can be connected directly to each HPLC system and used as 'ready to use' column.
This structure is a property of a new 'Sol-Gel' technology and consists of macropores and mesopores. The mesopores with an average diameter of 12 nm, form the fine porous structure of the column interior and create a very large surface area on which adsorption of the target compounds occurs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty