Would you like to log in again?
Pierce I-PER™, Insect Cell Protein Extraction Reagent, Thermo Scientific
Supplier: Thermo Scientific
Pierce
89802 89802
PI89802EA 412.25 USD InStock
PI89802 CAPI89802
Pierce I-PER™, Insect Cell Protein Extraction Reagent, Thermo Scientific
Buffers Extraction Buffers
I-PER™ Insect Cell Protein Extraction Reagent enables gentle and effective extraction of soluble protein from baculoviral-infected insect cells grown in suspension or monolayer culture.
  • Gentle extraction - optimized, mild non-ionic detergent provides maximum extraction of soluble proteins from insect cells
  • Effective - provides better protein extraction than sonication
  • Compatible - downstream compatibility with Western blotting, 6xHis-tagged protein purification, protein assays and ion-exchange chromatography
  • Flexible - useful for protein extraction from suspended or adherent cultured insect cells
The baculoviral insect cell expression system is an efficient and popular system for production of recombinant (eukaryotic) proteins in cell culture. Proteins expressed in baculoviral systems can be used for structural analyses, biochemical assays and a variety of other applications. I-PER Reagent maintains functionality of extracted proteins and is directly compatible with downstream applications such as protein assays, Western blotting and 6xHis-tagged protein purification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty