Would you like to log in again?
Plunging Tweezers for CP3 Cryoplunge 3, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CP3690
103306-160PK 281.13 USD InStock
103306-160
Plunging Tweezers for CP3 Cryoplunge 3, Electron Microscopy Sciences
Forceps
Custom tweezers specifically made to fit the Gatan Cryoplunge™ an instrument used in the preparation of frozen hydrated specimens for cryoEM.
  • High quality
  • Delicate handling of frozen hydated specimens
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty