Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human ACE Substrate 2

Supplier: AnaSpec
AS-60757
103006-002EA 247 USD InStock
103006-002
Human ACE Substrate 2
Proteins and Peptides
An ACE-2 (Angiotensin I-converting enzyme 2) fluorescent substrate. Complete hydrolysis of 0.04 mM results in a 300-fold fluorescence increase over background. Max Abs/Em=325/393 nm upon cleavage of substrate.
Sequence: Mca-APK(Dnp)
MW: 696.7 Da
% Peak area by HPLC: 95
Storage condition: -20°C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty