Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Ang III Peptide

Supplier: AnaSpec
AS-60704
103003-354EA 95 USD InStock
103003-354
Human Ang III Peptide
Proteins and Peptides
Angiotensin (Ang) III, RVYIHPF, is derived from the N-terminal cleavage of AngII, DRVYIHPF, through the action of aminopeptidase A (APA). AngIII is the main effector in the brain renin-angiotensin system (RAS) for vasopressin release.
Sequence: RVYIHPF
MW: 931.1 Da
% Peak area by HPLC: 95
Storage condition: -20°C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty