Would you like to log in again?
CUNO™ CTG-Klean System Filter Pack with Betapure™ BK Series Filter Cartridge
Supplier: 3M Healthcare
CUNO™
7GPK1BKZ8100G
76031-475CS 1365.12 USD InStock
76031-475
CUNO™ CTG-Klean System Filter Pack with Betapure™ BK Series Filter Cartridge
Filters Liquid Process Filters
CUNO™ CTG-Klean System Filter Pack with Betapure™ AU Series Cartridge as part of a totally enclosed system that uses a separate pressure vessel and disposable filter pack to isolate the product from the housing. It reduces the extensive time, labor and operator exposure to impurities and cleaning solvents involved with filter change-outs and cleaning of conventional housings.
  • Cartridge pack design reduces the extensive time and labor involved with filter change-outs and cleaning of conventional housings
  • Totally enclosed filter pack avoids handling of used filter cartridges
  • Design reduces gaskets, posts, spring & seal assemblies, and seal plates for efficient, cost- effective filtration
  • Filter pack can be capped and reused, depending on the application
  • Rapid filter change-out reduces labor time and the need for duplex systems
  • Betapure AU series filter cartridge’s rigid structure provides consistent and reproducible contaminant reduction
  • Betapure AU series filter cartridge provides excellent reduction of deformable contaminants for consistent effluent quality
CUNO™ CTG-Klean System was designed with versatility in mind to accommodate a wide variety of filter media, and offers compatibility with a range of fluids to meet a variety of process requirements.
Our line of Betapure™ AU series filters are state-of-the-art rigid, bicomponent fiber depth filters, available in absolute ratings from 2 to 190 microns. We manufacture them using our advanced fiber technology to provide the ultimate in consistent classifying filtration performance. The bonded rigid structure is non-deformable to withstand high differential pressure, fiber shedding, and to reduce media migration and provide absolute filtration - batch to batch. Betapure™ AU Series Filter Applications include Coatings - High Quality Paint, Film Coatings, Resins, and Ink, Food and Beverage - Bottled Water, Pre-Ro, Blend Water, Wash Water, General Industrial - Desalination, Plating, Machine Tool Coolant, Process Water, Electronics - Pre-Ro, Wafer Slurries, Ceramic Slurries, Chemical Mechanical Polishing, Cathode Ray Tube Production, Disc Cleaning and Chemical/Petro-Chemical - Process Water, Pre-Ro, Amine, Fine Chemicals.
The Rigid Construction Advantage.
To meet demanding filtration quality standards in today's market, absolute ratings will provide you with product consistency, and improve yields.
Lower pressure drops mean increased filter life, product throughput, and permit the use of fewer filters to achieve a given flow vs. differential pressure. When appropriately sized and rated, Betapure AU series filter cartridges employed in the CTG-Klean™ system work to meet your application specifications.
Go with Our Flow
Our cartridge pack design reduces the extensive time and labor involved with filter change-outs and cleaning of conventional housings, thereby reducing operator exposure to product cleaning solvents. This streamlined system, coupled with high performance, long-life filters, make CUNO CTG-Klean Systems the clear choice for cost-effective filtration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty