Would you like to log in again?
3M™ ScaleGard™ Pro Hardness Reduction Systems for Espresso Applications featuring Valve-in-Head Design, SGP195BN-T, 5633301
Supplier: 3M Healthcare
3M™
7100039092
76028-710EA 1072.76 USD InStock
76028-710
3M™ ScaleGard™ Pro Hardness Reduction Systems for Espresso Applications featuring Valve-in-Head Design, SGP195BN-T, 5633301
Filters Filter Cartridges
3M™ ScaleGard™ Pro Hardness Reduction System for Espresso Applications provides consistent quality water by reducing chlorine taste and odor. Built-in scale inhibition can help to prevent the potentially harmful effects of scale build-up on equipment. This system features a valve-in-head design.
  • Reduces calcium and magnesium, which create scale build-up
  • Reduces chlorine taste and odor
  • Multi-stage filtration reduces the need for a separate prefilter
  • Sanitary Quick Change (SQC) design makes cartridge change-outs easy without a filter wrench
Scale build-up has the potential to lead to increased maintenance costs and downtime. The Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench. This system features a valve-in-head design and is made for use in espresso applications.
Caution: Please note capacity in specifications is at 11 gpg hardness (180 mg/L hardness).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty