Would you like to log in again?
3M™ LifeASSURE™ BDA Series Filter Cartridge
Supplier: 3M Healthcare
LifeASSURE™
7010381548 7000050945 7000050939 7000051169 7010301285 7000050940
76029-717CS 7096.86 USD InStock
76029-717 76029-719 76029-721 76029-723 76029-725 76029-715
3M™ LifeASSURE™ BDA Series Filter Cartridge
Filters Filter Cartridges
PES, dual-layer, asymmetric microporous membrane filters for sterile filtration in food and beverage applications.
LifeASSURE BDA020 series filters contain two layers of PES membrane. The upstream layer is rated at 0.65 micron while the downstream layer is rated at 0.2 micron. Testing has shown this arrangement results in long service life while maintaining the necessary retention of microorganisms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty