Would you like to log in again?
3M™ Hot Water Softener for Commercial Warewashing Applications, Model HWS050
Supplier: 3M Healthcare
3M™
7000125570
76031-290EA 6764.22 USD InStock
76031-290
3M™ Hot Water Softener for Commercial Warewashing Applications, Model HWS050
Water Purification Systems Deionizers
3M™ Water Filtration Products HWS Series of warewashing water softener systems are designed to reduce spotting on dishes, glasses and flatware, as well as to reduce scale build-up on dishwasher nozzles and compartments
  • Space saving design allows unit to fit directly under dishwasher table for convenient storage
  • Corrosion resistant vinylester resin tank offers reduced deterioration and long term performance for temperatures up to 65.6 °C (150 °F)
  • Safety brine valve ensures overflow prevention in the event of a power outage
  • 12 day timer controlled regeneration allows system to clean and regenerate automatically without daily monitoring or interruption of service
The softened water also reduces hard scale build-up on dish machine walls and nozzles, which can reduce performance and equipment life. Softened water also allows detergents to work more efficiently, reducing excess chemical consumption.
A high capacity, high temperature resistant resin provides the ability to reduce calcium and magnesium minerals that cause white spots on dishware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty