Would you like to log in again?
3M™ High Flow Series System for Cold Beverage Applications, Model DWS160-L, 5627201
Supplier: 3M Healthcare
3M™
7100037914
76031-708EA 1008.01 USD InStock
76031-708
3M™ High Flow Series System for Cold Beverage Applications, Model DWS160-L, 5627201
Filters Filter Cartridges
The DWS160-L Water Filtration System is designed to provide consistent quality carbonated and non-carbonated beverages systems.
  • Provides consistent high-quality, great tasting water
  • Low-lead compliant materials
  • Sanitary Quick Change (SQC) encapsulated cartridge
This system helps to improve the taste, appearance and consistency of beverages at flow rates up to 2.5 gpm (9.4 lpm). Revolutionary Integrated Membrane Pre-Activated Carbon Technology (“IMPACT”) dual-zone media cartridge construction combines a membrane in series with premium activated carbon to provide superior throughput and cartridge life.
Certifications: NSF Certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty