Would you like to log in again?
3M™ High Flow Series System for Brewed Coffee & Hot Tea Applications featuring Valve-in-Head Design, Model BREW120-MS, 5616001
Supplier: 3M Healthcare
3M™
7000125417
76030-423CS 3245.05 USD InStock
76030-423
3M™ High Flow Series System for Brewed Coffee & Hot Tea Applications featuring Valve-in-Head Design, Model BREW120-MS, 5616001
Filters Filter Cartridges
3M™ High Flow Series System for Brewed Coffee & Hot Tea Applications featuring Valve–in–head Design, Model BREW120 – MS provides consistent high quality water by reducing particulate, chlorine taste and odor, cyst*, and scale**. The Sanitary Quick Change (SQC) design allows for easy cartridge change–outs.
  • Provides consistent high quality water
  • Reduces particulate, chlorine taste and odor, and cyst* from incoming water
  • Built–in scale inhibition can help to prevent the potentially harmful effects of scale build – up on equipment
  • Valve–in–head design
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • System designed for use in coffee and hot tea brewing applications
  • Includes replacement cartridge Model HF20–MS
Our 3M™ High Flow Series System for Brewed Coffee & Hot Tea Applications featuring Valve–in–head Design, Model BREW120–MS provides consistent high quality water by reducing particulate, chlorine taste and odor, cyst*, and scale**
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you're giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point–of–entry to dedicated point–of–use water filtration systems, look to 3M Purification to help you find the right solution.
*Based on the use of cryptosporidium parvum oocysts. **As tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty