Would you like to log in again?
3M™ High Flow Series Chloramines System for Cold Beverage Applications, Model HF195-CL, 5625802
Supplier: 3M Healthcare
3M™
7100074071
76030-498EA 1107.02 USD InStock
76030-498
3M™ High Flow Series Chloramines System for Cold Beverage Applications, Model HF195-CL, 5625802
Filters Filter Cartridges
3M™ High flow series chloramines system for cold beverage applications, Model HF195–CL provides consistent quality water by reducing particulate*, chloramines, and chlorine taste and odor.
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • Designed for chloramine reduction
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty