Would you like to log in again?
3M™ 1B Series Filter Housing
Supplier: 3M Healthcare
3M™
7000125595 7000125592 7000125594 7100031447 7000125593 7100058026
76031-798CS 2053.31 USD InStock
76031-798 76031-800 76028-922 76028-920 76031-796 76034-546
3M™ 1B Series Filter Housing
Filters Filter Cartridges
3M™ 1B series filter housing for general industrial filtration needs. Three-piece housings featuring center post construction, offer a choice of either cast iron and steel or 304 stainless steel as the materials construction. This 3M™ 1B series filter housing has flow rates of up to 16 gpm, and accepts working pressures and temperatures up to 175 psi at 250 °F.
  • Rugged cast iron and steel or long-wearing 304 stainless steel provides broad process compatibility
  • Mounting Pads Drilled and Tapped on housing head are easily installed using optional bracket
  • Design-Three-piece housing offers quick, clean, and convenient cartridge change-out
  • Center post construction for fast cartridge change-out
The 3M™ 1B series filter housing is available as one- or two-high cartridge models, and all models are available with or without a cartridge bypass valve (30 psid).
Applicable filter cartridges: Betapure™ AU series, Betafine™ series, 3M™ activated carbon series, Micro-Klean™ D series, Micro-Klean™ RB series.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty