Would you like to log in again?
3M™ OCS System for Office Coffee Applications featuring VH3 Valve-in-Head, Model VH3/HC350-S, 5626207
Supplier: 3M Healthcare
3M™
7100163126
76030-523CS 890.77 USD InStock
76030-523
3M™ OCS System for Office Coffee Applications featuring VH3 Valve-in-Head, Model VH3/HC350-S, 5626207
Filters Filter Cartridges
3M™ Water Filtration Products OCS System for Office Coffee Applications featuring VH3 Valve-in-Head, Cyst and Scale Reduction provides you with consistent quality water for hot beverages that can help increase repeat business and help to protect your equipment from hard scale build-up.
  • Provides consistent quality water for office coffee and hot cup applications by reducing sediment,* cysts, chlorine taste and odor
  • Reduces the ability of calcium and magnesium to form as hard scale* helping to reduce downtime and unscheduled service maintenance
  • Reduction of sediment* can help prevent equipment wear
  • Built-in valve allows cartridge change-outs without shutting off the incoming water supply, no external shut-off valve required
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
Our 3M™ Water Filtration Products OCS System for Office Coffee Applications featuring VH3 Valve-in-Head, Cyst and Scale Reduction for brewed coffee, tea and hot chocolate is designed to be effective at reducing sediment*, cysts, chlorine taste and odor, and scale* allowing you to feel more confident in the quality of the products you serve and can provide equipment protection from scale build-up
The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change-outs that are sanitary by design - requiring no contact with the filter media during cartridge change-out. The ramped interconnect completely disconnects the cartridge from the filter head with only a /14; turn, making this an extremely easy cartridge change-out.
Creative Filtration Technology
3M™ Water Filtration Products replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
*Tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty