Would you like to log in again?
3M™ OCS Vending Replacement Cartridge, Model VEN350-K, 5626105
Supplier: 3M Healthcare
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
3M™
7100041823
76030-509CS 625.28 USD InStock
76030-509
3M™ OCS Vending Replacement Cartridge, Model VEN350-K, 5626105
Filters Filter Cartridges
3M™ water filtration products model VEN350-K OCS vending replacement cartridge provides consistent high quality water for office vending applications by reducing sediment*, cyst**, asbestos and chlorine taste and odor. Scale inhibition helps to protect equipment from the potentially harmful effects of scale build-up. The Sanitary Quick Change (SQC) design allows for easy cartridge change-outs.
  • Provides consistent high quality water by reducing sediment*, cyst**, asbestos and chlorine taste and odor from incoming water
  • Built-in scale inhibitor helps to protect equipment from the potentially harmful effects of scale build-up
  • KDF® bacteriostatic media can provide additional protection for equipment
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench
The 3M™ water filtration products model VEN350-K OCS vending replacement cartridge is designed to be effective at reducing sediment*, cyst**, asbestos, and chlorine taste and odor from incoming water, allowing you to feel more confident in the quality of your water.
The cartridge's built-in scale inhibitor helps to protect equipment from the potentially harmful effects of scale build-up. KDF® bacteriostatic media can provide additional protection for your equipment. The Sanitary Quick Change (SQC) encapsulated design allows for fast and easy cartridge change-outs without the need for a filter wrench.
3M™ water filtration products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
A History of innovation
*As tested and verified by manufacturer's laboratory.
*Based on the use of Cryptosporidium parvum oocysts.
KDF is a trademark of KDF Fluid Treatment Inc.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty