Would you like to log in again?
3M™ OCS Office Water Replacement Cartridge, Model CC351, 5626107
Supplier: 3M Healthcare
3M™
7010381781
76030-513CS 411.09 USD InStock
76030-513
3M™ OCS Office Water Replacement Cartridge, Model CC351, 5626107
Filters Filter Cartridges
3M™ water filtration products Model CC351 OCS office water replacement cartridge provides consistent quality water by reducing sediment* and chlorine taste and odor for drinking water applications. The Sanitary Quick Change (SQC) design allows for easy cartridge change-outs without the need for a filter wrench. These replacement cartridges are to be used in VH3/CC351 systems.
  • Provides consistent quality water by reducing sediment* and chlorine taste and odor for drinking water applications
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
  • Cartridge change-outs do not require a filter wrench
  • Replacement cartridge for VH3/CC351 system
Our 3M™ water filtration products Model CC351 OCS office water replacement cartridge is designed to be effective at reducing sediment* and chlorine taste and odor for drinking water applications, allowing you to feel more confident with the quality of your water.
The Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench. These replacement cartridges are to be used in VH3/CC351 filter systems.
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
A history of innovation
*As tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty