Would you like to log in again?
3M™ CFS-M RO Membrane for SGLP-075, 5598729
Supplier: 3M Healthcare
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
3M™
7100048971
76028-341CS 3530.77 USD InStock
76028-341
3M™ CFS-M RO Membrane for SGLP-075, 5598729
Filters Filter Cartridges
3M™ CFS–M RO membrane for SGLP–RO has a sanitary quick change (SQC) design allows for easy cartridge change–outs.
  • Provides consistent quality water
  • Reverse osmosis replacement membrane for TDS reduction in boiler less steamers, combi–ovens and flash steamers
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • Replacement RO membrane for model SGLP–RO reverse osmosis systems
It is designed to reduce total dissolved solids (TDS) in boiler less steamers, combi–ovens and flash steamers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty