Would you like to log in again?
3M™ Aqua-Pure™ Whole House Large Diameter Stainless Steel Water Filter Housing SS12 EPE-316L
Supplier: 3M Healthcare
Aqua-Pure™
7000051153
76031-630EA 7340.76 USD InStock
76031-630
3M™ Aqua-Pure™ Whole House Large Diameter Stainless Steel Water Filter Housing SS12 EPE-316L
Filters Filter Cartridges
3M™ Aqua–Pure™ Whole House Large Diameter Stainless Steel Water Filter Housing SS12 EPE–316L filters all incoming water for your entire home or light commercial application. The housing can be used on both chlorinated (municipal) and non–chlorinated (well water) systems. Our SS12–EPE–316L housing provides unrestricted flow of up to 96 gpm with 2" NPT inlet/outlet connections.
  • Reduces sediment and chlorine taste and odor (cartridge dependent)
  • Accepts 12 double open–ended style cartridges, such as AP110, AP124, AP117 and AP420
  • Flow rate up to 96 gallons per minute (gpm)/363.4 lpm flow rate meets the water pressure demands of most homes and light commercial applications
  • 316L electro–polished stainless–steel head and sump helps provide corrosion resistance
  • Band clamp and heavy–duty gasket provides excellent durability
This means that the housing design can be used on both chlorinated (municipal) and non–chlorinated (well water) systems and in many different applications. In addition, the SS12–EPE–316L housing provides unrestricted flow of up to 96 gpm/363.4 lpm with 2" NPT inlet/outlet connections. Some other noteworthy features include: Maximum of 150 psi/1034 kPa pressure, Maximum use temperature of 121 °C (250 °F), For use with various standard diameter drop–in style cartridges for sediment and chlorine taste and odor reduction
The stainless–steel housing assembly with 2" NPT inlet/outlet connections helps provide high flow rates of up to 96 gallons per minute (gpm) /363.4 liters per minute (lpm) to meet the needs of most residential and light commercial applications. The 316 L electro–polished housing with mirror–like finish is a highly corrosion resistant material. The heavy–duty band clamp and gasket provide maximum durability. In addition, this stainless–steel housing accepts 12 double open–ended style cartridges such as: AP110, AP124, AP117 or the AP420. Once installed, these reduce sediment, scale and chlorine taste and odor.
3M™ Aqua–Pure™ branded water filtration products have been addressing water quality issues for over 50 years. Our products offer a wide range of solutions designed to address many types of water quality concerns. Many 3M Aqua–Pure products have the advantage of being backed by third–party certifications, such as NSF and WQA. For these reasons, 3M™ Aqua–Pure™ is a household brand name and a known leader in filtration technology. We’re recognized for innovation, trust and quality around the world.
For more than a century, 3M has applied innovation and technology to improving our customers’ lives and supporting their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. The 3M brand represents consistency, superior quality and value across thousands of products. Our success begins with a culture of creative collaboration that inspires a never–ending stream of powerful technologies and product solutions that make life better. We apply our technologies – often in combination – to an endless array of real–world customer needs. Of course, all of this is made possible by the people of 3M, and their singular commitment to make life easier and better for people around the world. Quite simply, 3M is committed to providing innovative, cost–effective products that help protect people, equipment and our environment. 3M Purification Inc. (formerly CUNO Incorporated), provides filtration and separation solutions for residential, food service, science labs, industrial, marine and other applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty