Would you like to log in again?
3M™ Aqua-Pure™ Whole House Large Diameter Stainless Steel Water Filter Housing SS8 EPE-316L
Supplier: 3M Healthcare
Aqua-Pure™
7000051145
76031-628EA 6845.89 USD InStock
76031-628
3M™ Aqua-Pure™ Whole House Large Diameter Stainless Steel Water Filter Housing SS8 EPE-316L
Filters Filter Cartridges
3M™ Aqua–Pure™ Whole House Large Diameter Stainless Steel Water Filter Housing SS8 EPE–316 L together with water filter cartridge filters all incoming water throughout an entire home or light commercial application. The housing design allows for use on both chlorinated (municipal) and non–chlorinated (well water) systems.
  • Reduces sediment and chlorine taste and odor and scale (cartridge dependent)
  • Accepts 8 double open–ended style cartridges, such as models AP110, AP124, AP117 and AP420
  • Flow rate up to 64 gallons per minute (gpm)/242.3 lpm meets the water pressure demands of most homes and light commercial applications
  • 316L electro–polished stainless–steel head and housing helps provide corrosion resistance
  • Band clamp and heavy–duty gasket provides excellent durability
3M Aqua–Pure branded water filtration products have been addressing water quality issues for over 50 years. Our products offer a wide range of solutions designed to address many types of water quality concerns. Many 3M Aqua–Pure products have the advantage of being backed by third–party certifications, such as NSF and WQA. For these reasons, 3M is a household brand name and a known leader in filtration technology. We're recognized for innovation, trust and quality around the world. Cleaner and clearer drinking water in your home is both a pleasure and a reassurance that you're giving your family the best. With a water filtration system from 3M Purification, you can provide excellent filtered drinking water for your home or business. From whole home water filtration to drinking water systems, look to 3M Purification to help you find the right solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty