Would you like to log in again?
3M™ Aqua-Pure™ In-Line Water Filter System IL-IM-01
Supplier: 3M Healthcare
Aqua-Pure™
7000050772
76030-453CS 1008.1 USD InStock
76030-453
3M™ Aqua-Pure™ In-Line Water Filter System IL-IM-01
Filters Filter Cartridges
3M™ Aqua–Pure™ In–Line Water Filter System, IL–IM–01 is suitable for refrigerators, drinking fountains, ice makers, coffee machines and drink dispensers can deliver cleaner, clearer, better tasting water and ice cubes. The single cartridge system reduces sediment and chlorine taste and odor at flow rates up to 0.5 gpm (1.89 lpm). Easy installations courtesy of the ¼" push–in fitting design.
  • Reduces sediment and chlorine taste and odor for clearer and cleaner drinking water, as well as ice cubes
  • Inline design is ideal for refrigerators, drinking fountains, ice makers, coffee machines and drink dispensers
  • Rated at 5 micron* to help provide sediment reduction as well as chlorine taste and odor reduction for up to 2500 gallons/9465 liters at 0.5 gpm/1.89 lpm
  • System is NSF/ANSI Standard 42 listed
3M™ Aqua–Pure™ In–Line Water Filter System, IL–IM–01, is a single cartridge that helps reduce sediment, chlorine taste and odor which may be in your drinking water and ice cubes
Ideal for the refrigerator that does not have a built – in water filter, this system can also be used for drinking fountains, coffee machines and drink dispensers. It provides effective filtration for up to 2500 gallons (9464 liters) at 0.5 gpm flow rate. The cartridge, using standard ¼" inlet/outlet connections, is sanitary in design and requires no contact with the filter media during change – out. The cartridge change–out frequency is every six months or sooner if water quality issues reduce water flow rates.
Features include: 25 to 125 psi /172 to 862 kPa pressure range, 40 to 100 °F/ 4.4 to 38 °C temperature range. Cartridges are sanitary in design, requiring no contact with the filter media during cartridge change – out. Filter cartridges require no pre – activation. In many filtration applications, carbon is often the preferred media of choice to absorb certain impurities and one of the most powerful absorbents available. Carbon is highly effective due to the vast surface area contained within a small footprint. One pound of carbon contains a surface area of approximately 100 to 125 acres and can absorb an extremely wide range of substances which may be in drinking water.
Ordering information: *As tested and verified by manufacturer’s laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty