Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
3M™ Integrity Test Device, 100, 3M
Supplier: 3M
7100089394
76029-158CS 12660.39 USD InStock
76029-158
3M™ Integrity Test Device, 100, 3M
Tool Sets
3M™ Series 100 Automated Integrity Testing Device is a hand-held portable membrane filter integrity tester for the Beverage Industry.
  • This compact, battery powered system can be operated independently of any AC supply for up to 12 hours
  • Should the unit remain uncharged it can retain all data in it’s memory for up to 6 months
The unit is designed to perform a pressure hold integrity test on membrane filter cartridges. The microprocessor-driven 3M™ Series 100 Automated Hand-held Integrity Testing Device can be programmed with up to 19 different test protocols, allowing the user to store commonly used test parameters covering all housing and filter combinations in use. Each of the different protocols can be quickly accessed through a single key stroke, with up to 200 test results being automatically stored ready to be recalled and printed at a later date.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty