Would you like to log in again?
3M™ ScaleGard™ HP Reverse Osmosis Replacment Cartridge, Model HFRO 500, 5626903
Supplier: 3M Healthcare
ScaleGard™
7000051420
76031-704EA 2214.41 USD InStock
76031-704
3M™ ScaleGard™ HP Reverse Osmosis Replacment Cartridge, Model HFRO 500, 5626903
Filters Filter Cartridges
3M™ ScaleGard™ HP Reverse Osmosis Replacement Cartridge, Model HFRO 500 has a Sanitary Quick Change (SQC) design allows for easy cartridge change-outs.
  • Provides consistent quality water
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench
  • Replacement membrane for 3M™ ScaleGard™ HP Reverse Osmosis System
Model HFRO 500 is designed for use in our 3M™ ScaleGard™ HP reverse osmosis system.
Our Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench.
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Recipe Quality Water™
A History of Innovation
For more than a century, 3M has applied innovation and technology to improve our customer's lives and support their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. Across thousands of products, the 3M brand represents consistency, superior quality, and value. Our success begins with a culture of creative collaboration that inspires a never-ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies - often in combination - to an endless array of real-world customer needs. Of course, all of this is made possible by the people of 3M and our singular commitment to make life easier and better for people around the world. Toward that end, 3M is committed to providing innovative, cost-effective products that help protect people, equipment and our environment. 3M Purification Inc. (formerly CUNO Incorporated) provides filtration and separation solutions for residential, foodservice, science labs, industrial, marine and other applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty