Would you like to log in again?
3M™ OCS In-line System for Water Cooler Applications, Model OW1005-L, 5628204
Supplier: 3M Healthcare
3M™
7010303344
76030-535CS 987.61 USD InStock
76030-535
3M™ OCS In-line System for Water Cooler Applications, Model OW1005-L, 5628204
Filters Filter Cartridges
3M™ Water Filtration Products OCS In-line Lead & Cyst Reduction System for Water Cooler Applications provides you with consistent quality water for bottle-less water coolers. Our products can help provide cleaner water while helping to protect your equipment from the potentially harmful effects of sediment*, cyst**, and chlorine taste and odor.
  • Provides consistent quality water for bottle-less water coolers by reducing sediment* and chlorine taste and odor, cyst** and lead
  • Can reduce equipment failure caused by chlorine
  • Reduces sediment* to help prevent premature equipment wear
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
  • Compact design allows for installation in tight spaces
3M™ Water Filtration Products OCS In-line Lead & Cyst Reduction System for Water Cooler Applications are an inexpensive way to help protect your equipment while providing cleaner, clearer, better tasting water from your bottle-less water cooler
It can also help reduce lead. The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change-outs that are sanitary by design - requiring no contact with the filter media during cartridge change-out.
3M™ Water Filtration Products branded replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
*Tested and verified by manufacturer's laboratory.
**As tested with E. Coli ATCC (11229). Tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty