Would you like to log in again?
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9812ELX, 5631709
Supplier: 3M Healthcare
3M™
7100045638
76030-603CS 1262.09 USD InStock
76030-603
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9812ELX, 5631709
Filters Filter Cartridges
3M™ Water Filtration Products Model CFS9812ELX Retrofit 9000 Series Replacement Cartridge provides consistent high quality water for beverages by reducing undesirable sediment*, chlorine taste and odor, and cyst** from incoming water. This extended length retrofit filter cartridge is compatible with most Everpure® foodservice heads.
  • Provides consistent high quality water by reducing undesirable sediment*, chlorine taste and odor, and cyst** from incoming water supply
  • Enhances water clarity for visually appealing beverages
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
  • Various 3M™ Water Filtration Products 9000 Series retrofit cartridges are available to meet particular water quality concerns
  • Extended length replacement cartridge allows more media for increased gallon capacity as compared to our standard length cartridges
Our 3M™ Water Filtration Products Model CFS9812ELX Retrofit 9000 Series Replacement Cartridge is designed to be effective at reducing undesirable sediment*, chlorine taste and odor, and cyst** from incoming water, allowing you to feel more confident in the quality of the products that you serve.
The Sanitary Quick Change (SQC) encapsulated design allows for fast and easy cartridge change-outs without the need for a filter wrench. This filter cartridge is compatible with most Everpure® foodservice heads, and various 3M Water Filtration Products 9000 Series retrofit cartridges are available to meet particular water quality concerns.
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Recipe Quality Water™
A History of Innovation
For more than a century, 3M has applied innovation and technology to improve our customers' lives and support their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. Across thousands of products, the 3M brand represents consistency, superior quality, and value. Our success begins with a culture of creative collaboration that inspires a never ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies - often in combination - to an endless array of real-world customer needs. Of course, all of this is made possible by the people of 3M and our singular commitment to make life easier and better for people around the world. Toward that end, 3M is committed to providing innovative, cost-effective products that help protect people, equipment, and our environment. 3M Purification Inc. (formerly CUNO Incorporated) provides filtration and separation solutions for residential, foodservice, science labs, industrial, marine, and other applications.
*Tested and verified by manufacturer's laboratory.
**Based on the use of Cryptosporidium parvum oocsyts.
Everpure is a trademark of Pentair Filtration Solutions, LLC.
Caution: Based on the use of Cryptosporidium parvum oocysts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty